Learn More
NDEL1 Mouse anti-Canine, Human, Mouse, Rat, Clone: OTI1G7, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF503254
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes a thiol-activated peptidase that is phosphorylated in M phase of the cell cycle. Phosphorylation regulates the cell cycle-dependent distribution of this protein, with a fraction of the protein bound strongly to centrosomes in interphase and localized to mitotic spindles in early M phase. Overall, this protein plays a role in nervous system development. Alternate transcriptional splice variants, encoding different isoforms, have been characterized.Specifications
| NDEL1 | |
| Monoclonal | |
| Unconjugated | |
| NDEL1 | |
| EOPA, MITAP1, NDE1L1, NDE2, NUDEL | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 170845, 489495, 81565, 83431 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI1G7 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q78PB6, Q9ERR1, Q9GZM8 | |
| NDEL1 | |
| Full length human recombit protein of human NDEL1 produced in HEk293 cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.