missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDFIP1 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-82012MFV500
This item is not returnable.
View return policy
Description
NDFIP1 Polyclonal antibody specifically detects NDFIP1 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NDFIP1 | |
| Polyclonal | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry-Paraffin | |
| Breast cancer-associated protein SGA-1M, MGC10924, N4WBP5Putative NF-kappa-B-activating protein 164, Nedd4 family interacting protein 1, NEDD4 family-interacting protein 1, NEDD4 WW domain-binding protein 5, Putative MAPK-activating protein PM13, Putative NFKB and MAPK-activating protein | |
| NDFIP1 antibody was raised against a 19 amino acid synthetic peptide near the amino terminus of human NDFIP1. The immunogen is located within the first 50 amino acids of NDFIP1. Amino Acid Squence: EEESGEPEQAAGDAPPPYS | |
| 0.1 mL | |
| Signal Transduction | |
| 80762 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction