missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDOR1 Antibody, Biotin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98487B
This item is not returnable.
View return policy
Description
NDOR1 Polyclonal antibody specifically detects NDOR1 in Human samples. It is validated for Western Blot
Specifications
| NDOR1 | |
| Polyclonal | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 27158 | |
| Store at 4°C in the dark. | |
| IgG |
| Western Blot | |
| Biotin | |
| bA350O14.9, EC 1.6, MGC138148, NADPH dependent diflavin oxidoreductase 1, NADPH-dependent diflavin oxidoreductase 1, NADPH-dependent FMN and FAD containing oxidoreductase, NADPH-dependent FMN and FAD-containing oxidoreductase, Novel reductase 1, NR1NADPH dependent FMN and FAD containing oxidoreductase | |
| Produced in rabbits immunized with E. coli-derived Human NDOR1 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction