missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDOR1 Antibody (OTI1E3), Alexa Fluor™ 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72896AF350
This item is not returnable.
View return policy
Description
NDOR1 Monoclonal specifically detects NDOR1 in Human, Mouse, Canine samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| NDOR1 | |
| Monoclonal | |
| Alexa Fluor 350 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| NDOR1 | |
| Full length recombinant protein of human NDOR1 (NP_055249) produced in HEK293T cells. | |
| 0.1 mL | |
| 27158.0 | |
| Store at 4°C in the dark. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI1E3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| bA350O14.9, EC 1.6, MGC138148, NADPH dependent diflavin oxidoreductase 1, NADPH-dependent diflavin oxidoreductase 1, NADPH-dependent FMN and FAD containing oxidoreductase, NADPH-dependent FMN and FAD-containing oxidoreductase, Novel reductase 1, NR1NADPH dependent FMN and FAD containing oxidoreductase | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Canine | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction