Learn More
NDOR1 Mouse anti-Canine, Human, Mouse, Clone: OTI1E3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF505464
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
This gene encodes an NADPH-dependent diflavin reductase that contains both flavin mononucleotide (FMN) and flavin adenine dinucleotide (FAD) binding domains. The encoded protein is an enzyme that catalyzes the transfers electrons from NADPH through FAD and FMN cofactors to potential redox partners. Alternative splicing results in multiple transcript variants.Specifications
| NDOR1 | |
| Monoclonal | |
| Unconjugated | |
| NDOR1 | |
| NR1, bA350O14.9 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 27158, 491234, 78797 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| OTI1E3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| A2AI05, Q9UHB4 | |
| NDOR1 | |
| Full length human recombit protein of human NDOR1 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG2a |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.