missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDP52 Antibody (OTI4H5), PE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72897PE
This item is not returnable.
View return policy
Description
NDP52 Monoclonal specifically detects NDP52 in Human, Monkey samples. It is validated for Flow Cytometry.
Specifications
| NDP52 | |
| Monoclonal | |
| PE | |
| PBS with 0.05% Sodium Azide | |
| CALCOCO2 | |
| Full length recombinant protein of human NDP52 (NP_005822) produced in HEK293T cells. | |
| 0.1 mL | |
| 10241.0 | |
| Store at 4°C in the dark. |
| Flow Cytometry | |
| OTI4H5 | |
| Flow Cytometry | |
| Antigen nuclear dot 52 kDa protein, calcium binding and coiled-coil domain 2, calcium-binding and coiled-coil domain-containing protein 2, NDP52MGC17318, Nuclear domain 10 protein 52, Nuclear domain 10 protein NDP52, Nuclear dot protein 52 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Monkey | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction