missing translation for 'onlineSavingsMsg'
Learn More

NDRG2 Antibody [Alexa Fluor« 647], Novus Biologicals Biologicals™

Product Code. 30504252 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30504252 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30504252 Supplier Novus Biologicals Supplier No. NBP338135AF647

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

NDRG2 Polyclonal antibody specifically detects NDRG2 in Human,Mouse samples. It is validated for ELISA,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDRG2
Applications ELISA, Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 647
Formulation 50mM Sodium Borate
Gene Alias DKFZp781G1938, FLJ25522, KIAA1248cytoplasmic protein Ndr1, NDR1-related protein NDR2, NDRG family member 2, protein NDRG2, Protein Syld709613, syld709613 protein, SYLDN-myc downstream regulator 2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 138-357 of human NDRG2 (NP_963832.1).,, Sequence:, LARYALNHPDTVEGLVLINIDPNAKGWMDWAAHKLTGLTSSIPEMILGHLFSQEELSGNSELIQKYRNIITHAPNLDNIELYWNSYNNRRDLNFERGGDITLRCPVMLVVGDQAPHEDAVVECNSKLDPTQTSFLKMADSGGQPQLTQPGKLTEAFKYFLQGMGYMASSCMTRLSRSRTASLTSAASVDGNRSRSRTLSQSSESGTLSSGPPGHTMEVSC
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 57447
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.