missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDRG4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA5144718

Product Code. 18085831

  • € 604.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: RQQIGNVVNQ ANLQLFWNMY NSRRDLDINR PGTVPNAKTL RCPVMLVVGD NAPAEDGVVE CNSKLDPTTT TFLKMADSGG LP Antigen sequence identity: Mouse 98%; Rat 98%.

This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signaling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NDRG4
Polyclonal
Unconjugated
NDRG4
BDM1; Brain development-related molecule 1; D8Bwg1337e; development-related protein; DKFZp686I1615; FLJ30586; FLJ42011; KIAA1180; MGC19632; Ndr1-rs; Ndr4; NDRG family member 4; NDRG4; N-myc downstream regulated 1, related sequence; N-myc downstream regulated 4; N-myc downstream regulated gene 4; N-myc downstream-regulated gene 4 protein; Protein Ndr4; protein NDRG4; R74996; smap8; SMAP-8; smooth muscle-associated protein 8; vascular smooth muscle cell-associated protein 8
Rabbit
Affinity chromatography
RUO
65009
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
IgG
Western Blot, Immunocytochemistry
0.05 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9ULP0
NDRG4
Recombinant protein fragment of Human NDRG4 (Product #RP-90840)
100 μL
Primary
Human
Antibody
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.