missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDRG4 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15664025
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15664025 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15664025 Supplier Invitrogen™ Supplier No. PA550095

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody detects endogenous levels of total NDRG4 protein.

This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signaling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDRG4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene NDRG4
Gene Accession No. Q8BTG7, Q9ULP0, Q9Z2L9
Gene Alias BDM1; Brain development-related molecule 1; D8Bwg1337e; development-related protein; DKFZp686I1615; FLJ30586; FLJ42011; KIAA1180; MGC19632; Ndr1-rs; Ndr4; NDRG family member 4; NDRG4; N-myc downstream regulated 1, related sequence; N-myc downstream regulated 4; N-myc downstream regulated gene 4; N-myc downstream-regulated gene 4 protein; Protein Ndr4; protein NDRG4; R74996; smap8; SMAP-8; smooth muscle-associated protein 8; vascular smooth muscle cell-associated protein 8
Gene Symbols NDRG4
Host Species Rabbit
Immunogen Synthesized peptide derived from internal of human NDRG4.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 234593, 64457, 65009
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.