missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDRG4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553299
This item is not returnable.
View return policy
Description
Immunogen sequence: RQQIGNVVNQ ANLQLFWNMY NSRRDLDINR PGTVPNAKTL RCPVMLVVGD NAPAEDGVVE CNSKLDPTTT TFLKMADSGG LP Highest antigen sequence identity to the following orthologs: Mouse - 98%, Rat - 98%.
This gene is a member of the N-myc downregulated gene family which belongs to the alpha/beta hydrolase superfamily. The protein encoded by this gene is a cytoplasmic protein that may be involved in the regulation of mitogenic signaling in vascular smooth muscles cells. Several alternatively spliced transcript variants of this gene have been described, but the full-length nature of some of these variants has not been determined.
Specifications
| NDRG4 | |
| Polyclonal | |
| Unconjugated | |
| NDRG4 | |
| BDM1; Brain development-related molecule 1; D8Bwg1337e; development-related protein; DKFZp686I1615; FLJ30586; FLJ42011; KIAA1180; MGC19632; Ndr1-rs; Ndr4; NDRG family member 4; NDRG4; N-myc downstream regulated 1, related sequence; N-myc downstream regulated 4; N-myc downstream regulated gene 4; N-myc downstream-regulated gene 4 protein; Protein Ndr4; protein NDRG4; R74996; smap8; SMAP-8; smooth muscle-associated protein 8; vascular smooth muscle cell-associated protein 8 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 65009 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9ULP0 | |
| NDRG4 | |
| Recombinant protein corresponding to Human NDRG4. Recombinant protein control fragment (Product #RP-90840). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction