missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDST3, Mouse anti-Human, Clone: 5B9, Abnova™
Mouse Monoclonal Antibody
Brand: Abnova H00009348-M01.100ug
This item is not returnable.
View return policy
Description
Sequence: IGFHKTSEKSVQSFQLKGFPFSIYGNLAVKDCCINPHSPLIRVTKSSKLEKGSLPGTDWTVFQINHSAYQPVIFAKVKTPENLSPSISKGAFYATIIHSpecifications
| NDST3 | |
| Monoclonal | |
| Unconjugated | |
| N-deacetylase/N-sulfotransferase (heparan glucosaminyl) 3 | |
| HSST3/MGC130028/MGC130029 | |
| Mouse | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody |
| ELISA, Western Blot | |
| 5B9 | |
| In 1x PBS, pH 7.4 | |
| NM_004784 | |
| NDST3 | |
| NDST3 (NP_004775, 162 a.a. ∼ 259 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. | |
| RUO | |
| 9348 | |
| Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction