Learn More
Invitrogen™ NDUFA1 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA550460
Product Code. 15694385
  • € 522.00 / 100µL
Description
The antibody detects endogenous levels of total NDUFA1 protein.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
NDUFA1
Polyclonal
Unconjugated
NDUFA1
1810049F12Rik; CI-MWFE; complex I MWFE subunit; complex I-MWFE; MWFE; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1 (7.5kD, MWFE); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 1, 7.5kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 1; NADH dehydrogenase 1 alpha subcomplex, 1; NADH oxidoreductase subunit MWFE; NADH ubiquinone oxidoreductase subunit MWFE; NADH:ubiquinone oxidoreductase (complex 1); NADH:ubiquinone oxidoreductase subunit A1; NADH-ubiquinone oxidoreductase MWFE subunit; NDUFA1; RGD1560955; type I dehydrogenase; ZNF183
Rabbit
Antigen affinity chromatography
RUO
363441, 4694, 54405
-20°C
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.5 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O15239, O35683
NDUFA1
Full length fusion protein of Human NDUFA1.
100 μL
Primary
Human, Mouse, Rat