missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFA10 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16566091
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16566091 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16566091 Supplier Invitrogen™ Supplier No. PA587650

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: LRYGMWHFLL GDKASKRLTE RSRVITVDGN ICTGKGKLAK EIAEKLGFKH FPEAGIHYPD STTGDGKPLA TDYNGNCSLE KFYDDPRSND GNSYRLQSWL YSSRLLQYSD ALEHLLTTGQ GVVLERSIFS DFVFLEAMYN QGFIRKQCVD HYNEVKSVTI CDYLPPHLVI YIDVPVPEVQ RRIQKKGDPH EMKITSAYLQ DIENAYKKTF LPEMSEKCEV LQYSAREAQD SKKVVEDIEY LKFDKGPWLK QDNRTLYHLR LLVQD; Positive Samples: HepG2, HL-60, BT-474, Jurkat, Mouse liver, Mouse kidney, Rat liver; Cellular Location: Mitochondrion matrix.

The protein encoded by this gene is a component of 42 kDa complex I, the first enzyme complex in the electron transport chain of mitochondria. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. A mutation in this gene was found in an individual with Leigh syndrome.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFA10
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.97 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene NDUFA10
Gene Accession No. O95299, Q99LC3
Gene Alias 2900053E13Rik; CI-42k; CI-42KD; complex I 42kDa subunit; complex I-42kD; FLJ21923; MGC5103; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex 10; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 10, 42kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 10, mitochondrial; NADH:ubiquinone oxidoreductase subunit A10; NADH-ubiquinone oxidoreductase 42 kDa subunit; Ndufa10; ndufa10l
Gene Symbols NDUFA10
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 36-300 of human NDUFA10 (NP_0045351).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4705, 67273, 678759
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.