missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA11 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94097-0.02ml
This item is not returnable.
View return policy
Description
NDUFA11 Polyclonal antibody specifically detects NDUFA11 in Mouse samples. It is validated for Western Blot
Specifications
| NDUFA11 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| B14.7, CI-B14.7, complex I B14.7 subunit, Complex I-B14.7, FLJ76501, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 11, 14.7kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 11, NADH-ubiquinone oxidoreductase subunit B14.7 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NDUFA11 (NP_783313.1). MAPKVFRQYWDIPDGTDCHRKAYSTTSIASVAGLTAAAYRVTLNPPGTFLEGVAKVGQYTFTAAAVGAVFGLTTCISAHVREKPDDPLNYFLGGCAGGLT | |
| 0.02 mL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 126328 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction