missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-98531MFV500
This item is not returnable.
View return policy
Description
NDUFA2 Polyclonal antibody specifically detects NDUFA2 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NDUFA2 | |
| Polyclonal | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| mFluor Violet 500 SE | |
| B8, CD14, CIB8, CI-B8, complex I B8 subunit, Complex I-B8, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2 (8kD, B8), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2, NADH-ubiquinone oxidoreductase B8 subunit, NADH-ubiquinone oxidoreductase subunit CI-B8 | |
| Produced in rabbits immunized with E. coli-derived Human NDUFA2 fragment. | |
| 0.1 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4695 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction