missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFA2 Polyclonal Antibody
Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596946
This item is not returnable.
View return policy
Description
Immunogen sequence: MAAAAASRGV GAKLGLREIR IHLCQRSPGS QGVRDFIEKR YVELKKANPD LPILIRECSD VQPKLWARYA FGQETNVPLN NFSADQVTRA LENVL; Positive Samples: Mouse kidney, Mouse heart; Cellular Location: Matrix side, Mitochondrion inner membrane, Peripheral membrane protein.
The NDUFA2 gene encodes one of the accessory subunits of complex I, the first and largest complex of the mitochondrial respiratory chain.
Specifications
| NDUFA2 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA2 | |
| AV000592; B8; C1-B8; CCDC76; CD14; CIB8; CI-B8; complex I B8 subunit; complex I-B8; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 2, 8kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 2; NADH:ubiquinone oxidoreductase subunit A2; NADH-ubiquinone oxidoreductase B8 subunit; NADH-ubiquinone oxidoreductase subunit CI-B8; NDUFA2 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 17991, 291660, 4695 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 2.18 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O43678, Q9CQ75 | |
| NDUFA2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-95 of human NDUFA2 (NP_0024791). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction