missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFA3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA570571
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Pig: 100%; Rabbit: 100%; Rat: 93%; Zebrafish: 77%.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFA3 | |
| Polyclonal | |
| Unconjugated | |
| NDUFA3 | |
| 1010001M12Rik; 1700022J01Rik; B9; CI-B9; complex I B9 subunit; complex I-B9; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 3, 9kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 3; NADH:ubiquinone oxidoreductase subunit A3; NADH-ubiquinone oxidoreductase B9 subunit; ndufa3; zgc:100908 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 66091 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9CQ91 | |
| NDUFA3 | |
| synthetic peptide directed towards the C-terminal region of mouse NDUFA3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction