missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93390-0.1ml
This item is not returnable.
View return policy
Description
NDUFA4 Polyclonal antibody specifically detects NDUFA4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| NDUFA4 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:50-1:100, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CI-9k, CI-MLRQ, Complex I 9kDa subunit, Complex I-MLRQ, FLJ27440, MGC104422, MGC126843, MGC126845, MLRQ, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4 (9kD, MLRQ), NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 4, 9kDa, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 4, NADH-ubiquinone oxidoreductase MLRQ subunit | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-81 of human NDUFA4 (NP_002480.1). MLRQIIGQAKKHPSLIPLFVFIGTGATGATLYLLRLALFNPDVCWDRNNPEPWNKLGPNDQYKFYSVNVDYSKLKKERPDF | |
| 0.1 mL | |
| Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4697 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction