missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA7 Monoclonal Antibody (OTI2C4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502749
This item is not returnable.
View return policy
Description
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
| NDUFA7 | |
| Monoclonal | |
| 0.33 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O95182 | |
| NDUFA7 | |
| Full length human recombit protein of human NDUFA7 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI2C4 | |
| Unconjugated | |
| NDUFA7 | |
| B14.5a, CI-B14.5a | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4701 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction