missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFA7 Monoclonal Antibody (OTI2E4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502701S
This item is not returnable.
View return policy
Description
NDUFA7 Monoclonal antibody specifically detects NDUFA7 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| NDUFA7 | |
| Monoclonal | |
| 0.53 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9NPC3 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 57820 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Flow Cytometry, Immunocytochemistry, Western Blot | |
| OTI2E4 | |
| Unconjugated | |
| CCNB1IP1 | |
| 14.5kDa; 2400007M02Rik; B14.5a; CI-B14.5a; complex I B14.5a subunit; complex I-B14.5a; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7; NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7 (B14.5a); NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, 7, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex subunit 7; NADH:ubiquinone oxidoreductase subunit A7; NADH-ubiquinone oxidoreductase subunit B14.5a; Ndufa7 | |
| Full length human recombinant protein of human CCNB1IP1 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction