Learn More
NDUFA7 Mouse anti-Human, Clone: OTI2E3, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF502657
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.Specifications
| NDUFA7 | |
| Monoclonal | |
| Unconjugated | |
| NDUFA7 | |
| B14.5a, CI-B14.5a | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4701 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Western Blot | |
| OTI2E3 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| O95182 | |
| NDUFA7 | |
| Full length human recombit protein of human NDUFA7 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.