missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFAB1 Recombinant Rabbit Monoclonal Antibody (JE65-76)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544832
This item is not returnable.
View return policy
Description
NDUFAB1 Recombinant Monoclonal Antibody for Western Blot, IHC (P), Flow
Carrier of the growing fatty acid chain in fatty acid biosynthesis in mitochondria. Accessory and non-catalytic subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), which functions in the transfer of electrons from NADH to the respiratory chain.
Specifications
| NDUFAB1 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O14561 | |
| NDUFAB1 | |
| Recombinant protein within human NDUFAB1 aa 57-156/156. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
| JE65-76 | |
| Unconjugated | |
| NDUFAB1 | |
| 2210401F17Rik; 2310039H15Rik; 2610003B19Rik; 8kDa; 9130423F15Rik; ACP; Acyl carrier protein, mitochondrial; CI-SDAP; complex I SDAP subunit; FASN2A; mitochondrial acyl carrier protein; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1; NADH dehydrogenase (ubiquinone) 1, alpha/beta subcomplex, 1, 8kDa; NADH:ubiquinone oxidoreductase SDAP subunit; NADH:ubiquinone oxidoreductase subunit AB1; NADH-ubiquinone oxidoreductase 9.6 kDa subunit; NDUFAB1; SDAP | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4706 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction