missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF4 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 204.00 - € 481.00
Specifications
| Antigen | NDUFAF4 |
|---|---|
| Dilution | Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18603540
|
Novus Biologicals
NBP2-93264-0.02ml |
0.02 mL |
€ 204.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18655330
|
Novus Biologicals
NBP2-93264-0.1ml |
0.1 mL |
€ 481.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFAF4 Polyclonal antibody specifically detects NDUFAF4 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| NDUFAF4 | |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 29078 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| bA22L21.1, C6orf66chromosome 6 open reading frame 66, Hormone-regulated proliferation-associated protein of 20 kDa, hormone-regulated proliferation-associated protein, 20 kDa, HRPAP20HSPC125, My013, NADH dehydrogenase (ubiquinone) 1 alpha subcomplex, assembly factor 4, NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 4 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-170 of human NDUFAF4 (NP_054884.1). MGALVIRGIRNFNLENRAEREISKMKPSVAPRHPSTNSLLREQISLYPEVKGEIARKDEKLLSFLKDVYVDSKDPVSSLQVKAAETCQEPKEFRLPKDHHFDMINIKSIPKGKISIVEALTLLNNHKLFPETWTAEKIMQEYQLEQKDVNSLLKYFVTFEVEIFPPEDKK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title