missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF5 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 198.00 - € 468.00
Specifications
| Antigen | NDUFAF5 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
NDUFAF5 Polyclonal antibody specifically detects NDUFAF5 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| NDUFAF5 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| bA526K24.2, C20orf7, chromosome 20 open reading frame 7, dJ842G6.1, EC 2.1.1, EC 2.1.1.-, FLJ22324, MGC90272, NADH dehydrogenase (ubiquinone) complex I, assembly factor 5, probable methyltransferase C20orf7, mitochondrial | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 196-345 of human NDUFAF5 (NP_077025.2). ELRCSLQLAETEREGGFSPHISPFTAVNDLGHLLGRAGFNTLTVDTDEIQVNYPGMFELMEDLQGMGESNCAWNRKALLHRDTMLAAAAVYREMYRNEDGSVPATYQIYYMIGWKYHESQARPAERGSATVSFGELGKINNLMPPGKKSQ | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 79133 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title