missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF5 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | C20orf7 |
|---|---|
| Dilution | Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18412891
|
Novus Biologicals
NBP2-14400-25ul |
25ul |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18147951
|
Novus Biologicals
NBP2-14400 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFAF5 Polyclonal specifically detects NDUFAF5 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| C20orf7 | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 79133 | |
| This antibody was developed against a recombinant protein corresponding to the amino acids: VNDLPRALEQIHYILKPDGVFIGAMFGGDTLYELRCSLQLAETEREGGFSPHISPFTAVNDLGHLLGRAGFNTLTVDTDEIQVNYPGMFELMED | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| bA526K24.2, C20orf7, chromosome 20 open reading frame 7, dJ842G6.1, EC 2.1.1, EC 2.1.1.-, FLJ22324, MGC90272, NADH dehydrogenase (ubiquinone) complex I, assembly factor 5, probable methyltransferase C20orf7, mitochondrial | |
| NDUFAF5 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title