missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFAF5 Mouse anti-Human, Clone: OTI6A9, TrueMAB™, Origene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA809182S
This item is not returnable.
View return policy
Description
NDUFAF5 Monoclonal antibody specifically detects NDUFAF5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NDUFAF5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q5TEU4 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79133 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI6A9 | |
| Unconjugated | |
| NDUFAF5 | |
| 2310003L22Rik; arginine-hydroxylase NDUFAF5, mitochondrial; bA526K24.2; C13H20orf7; C20orf7; C24H20orf7; dJ842G6.1; NADH dehydrogenase (ubiquinone) complex I, assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; NADH dehydrogenase [ubiquinone] 1 alpha subcomplex assembly factor 5; arginine-hydroxylase NDUFAF5, mitochondrial; NADH:ubiquinone oxidoreductase complex assembly factor 5; ndufaf5; probable methyltransferase C20orf7 homolog, mitochondrial; probable methyltransferase C20orf7, mitochondrial; putative methyltransferase C20orf7, mitochondrial-like protein; putative methyltransferase NDUFAF5; RGD1309829; zgc:162919 | |
| Full length human recombinant protein of human NDUFAF5 produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG2b |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction