missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFB1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15962415
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15962415 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15962415 Supplier Invitrogen™ Supplier No. PA575890

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

NDUFB1 (NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1), also known as CI-MNLL (complex I-MNLL), CI-SGDH or NADH-ubiquinone oxidoreductase MNLL subunit, is a 58 amino acid single-pass membrane protein that localizes to the matrix side of the mitochondrial membrane. A member of the complex I NDUFB1 subunit family, NDUFB1 is encoded by a gene that maps to human chromosome 14q32.12. Chromosome 14 houses over 700 genes and comprises nearly 3.5% of the human genome. Chromosome 14 encodes the presinilin 1 (PSEN1) gene, which is one of the three key genes associated with the development of Alzheimer's disease (AD). The SERPINA1 gene is also located on chromosome 14 and, when defective, leads to the genetic disorder alpha1-antitrypsin deficiency, which is characterized by severe lung complications and liver dysfunction.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFB1
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFB1
Gene Accession No. O75438, P0DN34, P0DN35
Gene Alias CI-MNLL; CI-SGDH; complex I MNLL subunit; complex I-MNLL; MNLL; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 1, 7kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 1; NADH:ubiquinone oxidoreductase subunit B1; NADH-ubiquinone oxidoreductase MNLL subunit; Ndufb1
Gene Symbols NDUFB1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues in Human NDUFB1.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100912357, 110324555, 4707
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.