Learn More
NDUFB11 Monoclonal Antibody (OTI2E10), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand:  Origene Technologies TA807710
Encompass_Preferred
 View more versions of this product
Product Code. 17558423
  • € 540.87 / 100µL
Description
NDUFB11 is an accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I) that is believed not to be involved in catalysis. NDUFB11 functions in the transfer of electrons from NADH to the respiratory chain and is located in the inner mitochondrial membrane. The immediate electron acceptor for NDUFB11 is believed to be ubiquinone. Further, NDUFB11 is believed to have NADH dehydrogenase activity, and oxidoreductase activity. Diseases associated with NDUFB11 protein dysfunction include linear skin defects with multiple congenital anomalies and mitochondrial complex I deficiency.
Specifications
Specifications
NDUFB11
Monoclonal
1 mg/mL
PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Q9NX14
NDUFB11
Human recombit protein fragment corresponding to amino acids 30-163 of human NDUFB11 produced in E.coli.
100 μL
Primary
Human
Antibody
IgG1
Western Blot
OTI2E10
Unconjugated
NDUFB11
CI-ESSS, ESSS, NP17.3, Np15, P17.3
Mouse
Affinity Chromatography
RUO
54539
-20° C, Avoid Freeze/Thaw Cycles