missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB5 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97853UV
This item is not returnable.
View return policy
Description
NDUFB5 Polyclonal specifically detects NDUFB5 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| NDUFB5 | |
| Polyclonal | |
| Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| 5 (16kD, SGDH), CISGDH, CI-SGDH, Complex I-SGDH, DKFZp686N02262, FLJ30597, MGC111204, MGC12314, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial, SGDH | |
| Produced in rabbits immunized with E. coli-derived Human NDUFB5 fragment. | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| 4711 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction