missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: EGYVPEHWEY YKHPISRWIA RNFYDSPEKI YERTMAVLQI EAEKAELRVK ELEVRKLMHV RGDGPWYYYE TIDKELIDHS PKATPDN Highest antigen sequence identity to the following orthologs: Mouse - 79%, Rat - 77%.
NDUFB5 is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. The protein encoded by this gene is a subunit of the multisubunit NADH:ubiquinone oxidoreductase (complex I). Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
Specifications
| Antigen | NDUFB5 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | NDUFB5 |
| Gene Accession No. | O43674 |
| Gene Alias | 0610007D05Rik; 16 kDa precursor; AU015782; CISGDH; CI-SGDH; complex I SGDH subunit; complex I-SGDH; DKFZp686N02262; FLJ30597; hm:zeh0024; hypothetical protein LOC557853; MGC111204; MGC12314; NADH dehydrogenase; NADH dehydrogenase (ubiquinone) 1 beta subcomplex 5; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 5, 16kDa, precursor; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 5, mitochondrial; NADH dehydrogenase 1 beta subcomplex 5; NADH:ubiquinone oxidoreductase subunit B5; NADH-ubiquinone oxidoreductase SGDH subunit; Ndufb5; SGDH; zgc:123331 |
| Gene Symbols | NDUFB5 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
