missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB9 Antibody (OTI8B7), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72918MFV500
This item is not returnable.
View return policy
Description
NDUFB9 Monoclonal antibody specifically detects NDUFB9 in Human, Mouse, Rat, Primate, Monkey samples. It is validated for Flow Cytometry
Specifications
| NDUFB9 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Primate, Monkey | |
| Purified |
| Flow Cytometry | |
| OTI8B7 | |
| Flow Cytometry | |
| B22, CI-B22, complex I B22 subunit, Complex I-B22, FLJ22885, LYR motif-containing protein 3, LYRM3DKFZp566O173, NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa, NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9, NADH-ubiquinone oxidoreductase B22 subunit, UQOR22NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9 (22kD, B22) | |
| Human fragment corresponding to amino acids 3-179 of human NDUFB9 (NP_001893) produced in E.coli. | |
| 0.1 mL | |
| Endocrinology, Neurodegeneration, Neuroscience | |
| 4715 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction