missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB9 Monoclonal Antibody (OTI4F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502553S
This item is not returnable.
View return policy
Description
NDUFB9 Monoclonal antibody specifically detects NDUFB9 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| NDUFB9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q06413 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 4208 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F8 | |
| Unconjugated | |
| MEF2C | |
| 1190008J14Rik; B22; CI-B22; complex I B22 subunit; complex I-B22; EGK_19260; LYR motif-containing protein 3; LYRM3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9; NADH:ubiquinone oxidoreductase subunit B9; NADH-ubiquinone oxidoreductase B22 subunit; NDUFB9; UQOR22 | |
| Full length human recombinant protein of human MEF2C produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction