missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB9 Monoclonal Antibody (OTI4F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA502553
This item is not returnable.
View return policy
Description
LYRM3, also known as NADH dehydrogenase (ubiquinone) 1 beta subcomplex 9 (NDUFB9), is a ubiquitously expressed LYR-motif containing protein. It has been suggested to be a candidate gene for the branchio-oto-renal (BOR) syndrome, which is characterized by branchial and renal abnormalities and heredity deafness disorders. Other than its LYR-motif, LYRM3 appears to have no functional or structural relationship to either LYRM1 or LYRM2.Specifications
| NDUFB9 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9CQJ8, Q9Y6M9 | |
| Ndufb9 | |
| Human recombit protein fragment corresponding to amino acids 3-179 of human NDUFB9 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI4F8 | |
| Unconjugated | |
| NDUFB9 | |
| B22, CI-B22, LYRM3, UQOR22 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 299954, 4715, 475094, 66218 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction