missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFB9 Monoclonal Antibody (OTI8B7), TrueMAB™, OriGene
Description
NDUFB9 Monoclonal antibody specifically detects NDUFB9 in Human samples. It is validated for Flow Cytometry, Western Blot
Specifications
Specifications
| Antigen | NDUFB9 |
| Applications | Flow Cytometry, Western Blot |
| Classification | Monoclonal |
| Clone | OTI8B7 |
| Concentration | 0.76 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide |
| Gene | MEF2C |
| Gene Accession No. | Q06413 |
| Gene Alias | 1190008J14Rik; B22; CI-B22; complex I B22 subunit; complex I-B22; EGK_19260; LYR motif-containing protein 3; LYRM3; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9; NADH dehydrogenase (ubiquinone) 1 beta subcomplex, 9, 22kDa; NADH dehydrogenase [ubiquinone] 1 beta subcomplex subunit 9; NADH:ubiquinone oxidoreductase subunit B9; NADH-ubiquinone oxidoreductase B22 subunit; NDUFB9; UQOR22 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?