missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFC2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116609
This item is not returnable.
View return policy
Description
Positive Samples: 293T, OVCAR3, HeLa, A-549, mouse stomach, mouse brain, mouse heart, rat kidney Immunogen sequence: MIARRNPEPL RFLPDEARSL PPPKLTDPRL LYIGFLGYCS GLIDNLIRRR PIATAGLHRQ LLYITAFFFA GYYLVKREDY LYAVRDREMF GYMKLHPEDF.
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFC2 | |
| Polyclonal | |
| Unconjugated | |
| NDUFS2 | |
| 14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 1810004I06Rik; 2010300P09Rik; B14.5b; CI-B14.5b; CI-B145b; complex I subunit B14.5b; complex I-B14.5b; Complex I-B145b; G1; HLC1; HLC-1; Human lung cancer oncogene 1 protein; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 subunit C2; NADH:ubiquinone oxidoreductase subunit C2; NADHDH2; NADH-ubiquinone oxidoreductase subunit B14.5b; Ndufc2; R75094 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 226646, 289218, 293130, 4718, 4720, 68197 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.84 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| O75306, O95298, Q641Y2, Q91WD5, Q9CQ54 | |
| NDUFC2, NDUFS2 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human NDUFC2 (NP_0045401). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction