missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFC2 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15726451 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15726451 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15726451 Supplier Invitrogen™ Supplier No. PA558572

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: MIARRNPEPL RFLPDEARSL PPPKLTDPRL LYIGFLGYCS GLIDNLIRRR P Highest antigen sequence identity to the following orthologs: Mouse - 71%, Rat - 75%.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFC2
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene NDUFS2
Gene Accession No. O75306, O95298
Gene Alias 14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 1810004I06Rik; 2010300P09Rik; B14.5b; CI-B14.5b; CI-B145b; complex I subunit B14.5b; complex I-B14.5b; Complex I-B145b; G1; HLC1; HLC-1; Human lung cancer oncogene 1 protein; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 subunit C2; NADH:ubiquinone oxidoreductase subunit C2; NADHDH2; NADH-ubiquinone oxidoreductase subunit B14.5b; Ndufc2; R75094
Gene Symbols NDUFC2, NDUFS2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human NDUFC2. Recombinant protein control fragment (Product #RP-97411).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 4718, 4720
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.