missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFC2 Recombinant Rabbit Monoclonal Antibody (JE65-31)
GREENER_CHOICE

Rabbit Recombinant Monoclonal Antibody

Brand:  Invitrogen™ MA550672

Product Code. 17929412

  • € 502.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Positive control: 293T cell lysate, Hela cell lysate, A549 cell lysate, human kidney tissue lysate. Predicted band size: 14 kDa Subcellular Location: Mitochondrion inner membrane.

Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

NDUFC2
Recombinant Monoclonal
1 mg/mL
TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
O75306, O95298
NDUFC2, NDUFS2
Recombinant protein within human NDUFC2 aa 1-119/119.
100 μL
Primary
Human
Antibody
IgG
Western Blot
JE65-31
Unconjugated
NDUFS2
14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 1810004I06Rik; 2010300P09Rik; B14.5b; CI-B14.5b; CI-B145b; complex I subunit B14.5b; complex I-B14.5b; Complex I-B145b; G1; HLC1; HLC-1; Human lung cancer oncogene 1 protein; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 subunit C2; NADH:ubiquinone oxidoreductase subunit C2; NADHDH2; NADH-ubiquinone oxidoreductase subunit B14.5b; Ndufc2; R75094
Rabbit
Protein A
RUO
4718, 4720
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.