missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFC2 Recombinant Rabbit Monoclonal Antibody (JE65-31)

Rabbit Recombinant Monoclonal Antibody
Brand: Invitrogen™ MA544685
This item is not returnable.
View return policy
Description
NDUFC2 Recombinant Monoclonal Antibody for Western Blot, IHC (P)
Accessory subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase (Complex I), that is believed not to be involved in catalysis. Complex I functions in the transfer of electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone.
Specifications
| NDUFC2 | |
| Recombinant Monoclonal | |
| 1 mg/mL | |
| TBS with 0.05% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| O75306, O95298 | |
| NDUFC2, NDUFS2 | |
| Recombinant protein within human NDUFC2 aa 1-119. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Immunohistochemistry (Paraffin), Western Blot | |
| JE65-31 | |
| Unconjugated | |
| NDUFS2 | |
| 14.5b subunit of the NADH-ubiquinone dehydrogenase complex; 1810004I06Rik; 2010300P09Rik; B14.5b; CI-B14.5b; CI-B145b; complex I subunit B14.5b; complex I-B14.5b; Complex I-B145b; G1; HLC1; HLC-1; Human lung cancer oncogene 1 protein; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2; NADH dehydrogenase (ubiquinone) 1, subcomplex unknown, 2, 14.5kDa; NADH dehydrogenase [ubiquinone] 1 subunit C2; NADH:ubiquinone oxidoreductase subunit C2; NADHDH2; NADH-ubiquinone oxidoreductase subunit B14.5b; Ndufc2; R75094 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 4718, 4720 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction