missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NDUFS2 Monoclonal Antibody (OTI1D5)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA526536
This item is not returnable.
View return policy
Description
NDUFS2 Monoclonal Antibody for IHC (P), Western Blot
The protein encoded by this gene is a core subunit of the mitochondrial membrane respiratory chain NADH dehydrogenase. Mammalian mitochondrial complex I is composed of at least 43 different subunits, 7 of which are encoded by the mitochondrial genome, and the rest are the products of nuclear genes. The iron-sulfur protein fraction of complex I is made up of 7 subunits, including this gene product. Complex I catalyzes the NADH oxidation with concomitant ubiquinone reduction and proton ejection out of the mitochondria. Mutations in this gene are associated with mitochondrial complex I deficiency. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| NDUFS2 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol, 1% BSA and 0.02% sodium azide; pH 7.3 | |
| O75306 | |
| Ndufs2 | |
| Human recombinant protein fragment corresponding to amino acids 205-463 of NDUFS2 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1D5 | |
| Unconjugated | |
| Ndufs2 | |
| AL033311; CI-49; CI-49kD; comp; complex 1, mitochondrial respiratory chain, 49-KD subunit; complex I 49kDa subunit; complex I-49kD; EMBL:AAH82067.1}; NADH dehydrogenase (ubiquinone) Fe-S protein 2; NADH dehydrogenase (ubiquinone) Fe-S protein 2, 49kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 2, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S2; NADH-coenzyme Q reductase; NADH-ubiquinone oxidoreductase 49 kDa subunit; NADH-ubiquinone oxidoreductase NDUFS2 subunit; Ndufs2; ndufs2 {ECO:0000312; unnamed protein product | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 4720 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction