missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NDUFS3 Recombinant Rabbit Monoclonal Antibody (HL2097)

Product Code. 18075841
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18075841 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18075841 Supplier Invitrogen™ Supplier No. MA550381

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

NDUFS3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

This gene encodes one of the iron-sulfur protein components of mitochondrial NADH:ubiquinone oxidoreductase. Mutations in this gene are associated with Leigh syndrome resulting from mitochondrial complex I deficiency.
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS3
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone HL2097
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene NDUFS3
Gene Accession No. O75489, Q9DCT2
Gene Alias 0610010M09Rik; CI-30; CI-30kD; complex I 30kDa subunit; complex I-30kD; hypothetical protein LOC550451; NADH dehydrogenase (ubiquinone); NADH dehydrogenase (ubiquinone) Fe-S protein 3; NADH dehydrogenase (ubiquinone) Fe-S protein 3, (NADH-coenzyme Q reductase); NADH dehydrogenase (ubiquinone) Fe-S protein 3, 30kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 3, mitochondrial; NADH dehydrogenase-ubiquinone 30 kDa subunit; NADH:ubiquinone oxidoreductase core subunit S3; NADH-ubiquinone oxidoreductase 30 kDa subunit; NDUFS3; zgc:112520
Gene Symbols NDUFS3
Host Species Rabbit
Immunogen Recombinant fragment of human NDUFS3.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 295923, 4722, 550451, 68349
Target Species Human, Mouse, Rat, Zebrafish
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.