missing translation for 'onlineSavingsMsg'
Learn More

NDUFS5 Antibody (5C2) [mFluor Violet 610 SE], Novus Biologicals Biologicals™

Product Code. 30521167 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30521167 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30521167 Supplier Novus Biologicals Supplier No. NBP343006MFV610

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

NDUFS5 Monoclonal antibody specifically detects NDUFS5 in Human samples. It is validated for ELISA,Immunohistochemistry,Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS5
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 5C2
Conjugate mFluor Violet 610 SE
Formulation 50mM Sodium Borate
Gene Alias CI-15 kDa, CI-15k, CI15K, Complex I-15 kDa, NADH dehydrogenase (ubiquinone) Fe-S protein 5 (15kD) (NADH-coenzyme Qreductase), NADH dehydrogenase (ubiquinone) Fe-S protein 5, 15kDa (NADH-coenzyme Qreductase), NADH dehydrogenase [ubiquinone] iron-sulfur protein 5, NADH:ubiquinone oxidoreductase 15 kDa IP subunit, NADH-ubiquinone oxidoreductase 15 kDa subunit
Host Species Mouse
Immunogen NADH dehydrogenase(ubiquinone) Fe-S protein 5, 15kDa(NADH-coenzyme Q reductase) - (Uniprot# O43920)
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 4725
Target Species Human
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.