missing translation for 'onlineSavingsMsg'
Learn More

NDUFS8 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17895732
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17895732 100 μg 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17895732 Supplier Invitrogen Supplier No. PA5142594

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

NDUFS8 Polyclonal Antibody

NDUFS8 Polyclonal Antibody for Western Blot

TRUSTED_SUSTAINABILITY

Specifications

Antigen NDUFS8
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Gene NDUFS8
Gene Accession No. O00217, Q8K3J1
Gene Alias BC021616; CI-23k; CI23KD; CI-23kD; complex I 23kDa subunit; complex I-23kD; NADH dehydrogenase (ubiquinone) Fe-S protein 8; NADH dehydrogenase (ubiquinone) Fe-S protein 8, 23kDa (NADH-coenzyme Q reductase); NADH dehydrogenase [ubiquinone] iron-sulfur protein 8, mitochondrial; NADH:ubiquinone oxidoreductase core subunit S8; NADH-ubiquinone oxidoreductase 23 kDa subunit; NDUFS8; TYKY; TYKY subunit
Gene Symbols NDUFS8
Host Species Goat
Immunogen Peptide with sequence C-AEPRADGSRR.
Purification Method Ammonium Sulfate Precipitation
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 225887, 4728
Target Species Human, Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.