Learn More
NDUFV1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 11238-1-AP-150UL
 View more versions of this product
Product Code. 16808183
  • € 410.00 / 150µL
Description
NDUFV1 is the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.The NDUFV1 gene encodes the 51-kD subunit of complex I (NADH:ubiquinone oxidoreductase) of the mitochondrial respiratory chain.
Specifications
Specifications
NDUFV1
Polyclonal
Unconjugated
NDUFV1
CI 51kD, Complex I 51kD, NDUFV1, UQOR1
Rabbit
Antigen Affinity Chromatography
RUO
17995, 293655, 4723
-20°C
Liquid
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
0.3 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P49821, Q91YT0
Ndufv1
NDUFV1 Fusion Protein Ag1764
150 μL
Primary
Human, Mouse, Rat