missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV2 Antibody, Allophycocyanin, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99225APC
This item is not returnable.
View return policy
Description
NDUFV2 Polyclonal antibody specifically detects NDUFV2 in Human samples. It is validated for Western Blot
Specifications
| NDUFV2 | |
| Polyclonal | |
| Western Blot | |
| CI-24k, mitochondrial respitory chain, 24 kD subunit, NADH dehydrogenase (ubiquinone) flavoprotein 2 (24kD), NADH dehydrogenase (ubiquinone) flavoprotein 2, 24kDa, NADH dehydrogenase [ubiquinone] flavoprotein 2, mitochondrial, NADH dehydrogenase ubiquinone flavoprotein 2, mitochondrial, NADH-ubiquinone oxidoreductase 24 kDa subunit, NADH-ubiquinone oxidoreductase flavoprotein 2, nuclear-encoded mitochondrial NADH-ubiquinone reductase 24Kd subunit | |
| Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human NDUFV2. | |
| 0.1 mL | |
| metabolism | |
| 4729 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot | |
| APC | |
| PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction