missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NDUFV3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
€ 433.00 - € 572.00
Specifications
| Antigen | NDUFV3 |
|---|---|
| Dilution | Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18464622
|
Novus Biologicals
NBP1-85622-25ul |
25 μL |
€ 433.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18216106
|
Novus Biologicals
NBP1-85622 |
0.1 mL |
€ 572.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
NDUFV3 Polyclonal specifically detects NDUFV3 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| NDUFV3 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 4731 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:RVVSKGRGGLRKPEASHSFENRAPRVTVSAKEKTLLQKPHVDITDPEKPHQPKKKGSPAKPSEGRENARPKTTMPRSQVDEEFLKQSLKEKQLQKTFRLNEIDKESQKPFEVKGPLPVHTKSGLSA | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| CI-10k, CI-9KD, complex I 10kDa subunit, complex I, mitochondrial respiratory chain, 10-kD subunit, Complex I-9kD, mitochondrial NADH oxidoreductase-like protein, NADH dehydrogenase (ubiquinone) flavoprotein 3 (10kD), NADH dehydrogenase (ubiquinone) flavoprotein 3, 10kDa, NADH dehydrogenase [ubiquinone] flavoprotein 3, mitochondrial, NADH-ubiquinone oxidoreductase 9 kD subunit, NADH-ubiquinone oxidoreductase 9 kD subunit precursor, EC 1.6.5.310CI-9kD, NADH-ubiquinone oxidoreductase 9 kDa subunit, NADH-ubiquinone oxidoreductase flavoprotein 3, 10kD, Renal carcinoma antigen NY-REN-4 | |
| NDUFV3 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title