missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ NE 100 hydrochloride
GREENER_CHOICE

Product Code. 18094652
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18094652

Brand: Tocris Bioscience™ 3133/50

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective σ1 antagonist

Potent and selective σ1 receptor antagonist (Ki = 0.86 nM) that displays > 55-fold selectivity over σ2 receptors and > 6000-fold selectivity over D1, D2, 5-HT1A, 5-HT2 and PCP receptors. Exhibits reversible binding (Kd = 1.2 nM) and displays antipsychotic activity in vivo. Orally active.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 149409-57-4
Molecular Formula C23H34ClNO2

Specifications

Quantity 50 mg
Formula Weight 391.97
Purity >98%
Molecular Weight (g/mol) 391.98
Chemical Name or Material NE 100 hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.