Learn More
Memmert™ Nearly Gastight Interior with Gas Inlet/Outlet for UF450
Shop All Memmert Products
Click to view available options
For Use With (Equipment):
UF450 Oven
Unit Size:
Each
Description
- Gas inlet/outlet through 2 tubes with ball valves
- Technical clarification required for combination with other options
Specifications
Specifications
| For Use With (Equipment) | UF450 Oven |
| Type | Interior |
| Includes | Gas Inlet/Outlet |