missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAB1 Antibody (NECAB1/7677), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-20934JF646
This item is not returnable.
View return policy
Description
NECAB1 Monoclonal antibody specifically detects NECAB1 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| NECAB1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1 | |
| Recombinant human NECAB1 protein | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| NECAB1/7677 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 64168 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction