missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NECAB1 Antibody (OTI4G8), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72923V
This item is not returnable.
View return policy
Description
NECAB1 Monoclonal specifically detects NECAB1 in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry.
Specifications
| NECAB1 | |
| Monoclonal | |
| DyLight 405 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| NECAB1 | |
| Full length recombinant protein of human NECAB1 (NP_071746) produced in HEK293T cells. | |
| 0.1 mL | |
| 64168.0 | |
| Store at 4°C in the dark. |
| Western Blot, Flow Cytometry | |
| OTI4G8 | |
| Western Blot, Flow Cytometry | |
| EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1 | |
| Mouse | |
| Affinity Purified | |
| Primary | |
| Human, Mouse, Rat | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction