Learn More
NECAB1 Antibody (OTI4G8), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-72923MFV450
Product Code. 18457793
  • € 773.00 / 0.10mL
Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Description
Description
NECAB1 Monoclonal antibody specifically detects NECAB1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry
TRUSTED_SUSTAINABILITY
Spécification
Spécification
NECAB1
Monoclonal
mFluor Violet 450 SE
50mM Sodium Borate
Mouse
Immunogen affinity purified
RUO
64168
Store at 4C in the dark.
IgG2a
Flow Cytometry
OTI4G8
Flow Cytometry
EF hand calcium binding protein 1, EFCBP1, EF-hand calcium binding protein 1, EF-hand calcium-binding protein 1, neuronal calcium binding protein, Neuronal calcium-binding protein 1, N-terminal EF-hand calcium binding protein 1, N-terminal EF-hand calcium-binding protein 1, STIP-1, synaptotagmin interacting protein 1
Full length human recombinant protein of human NECAB1 (NP_071746) produced in HEK293T cell.
0.1 mL